Assay Details
| Gene Symbol: HTATIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015290397.2 | XP_015145883.1 | 2 - 3 | 335 | 69 | |
| XM_422206.6 | XP_422206.1 | 2 - 3 | 313 | 69 | ||
| GenBank mRNA | BX931233.1 | - | 2 - 3 | 265 | 69 | |
| BX933391.1 | - | 1 - 2 | 275 | 69 | ||
| BX935289.1 | - | 2 - 3 | 306 | 69 | ||
More Information
Set Membership: |
|

HTATIP2
Genomic Map