Assay Details
| Gene Symbol: RPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277164.1 | NP_001264093.1 | 4 - 5 | 373 | 129 | |
| GenBank mRNA | BX933609.1 | - | 3 - 4 | 359 | 129 | |
| BX934117.1 | - | 3 - 4 | 356 | 129 | ||
| BX934597.2 | - | 3 - 4 | 359 | 129 | ||
| BX935640.2 | - | 3 - 4 | 359 | 129 | ||
| CN226583.1 | - | 3 - 4 | 364 | 129 | ||
| CR386179.1 | - | 3 - 4 | 263 | 129 | ||
| CR386183.1 | - | 3 - 4 | 355 | 129 | ||
More Information
Set Membership: |
|

RPS2
Genomic Map