Assay Details
| Gene Symbol: TOMM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302134.1 | NP_001289063.1 | 155 | 78 | ||
| GenBank mRNA | BX930946.1 | - | 138 | 78 | ||
| BX933804.1 | - | 146 | 78 | |||
| BX933812.1 | - | 146 | 78 | |||
| BX950834.2 | - | 136 | 78 | |||
| CN231382.1 | - | 155 | 78 | |||
More Information
Set Membership: |
|

TOMM6
Genomic Map