Assay Details
| Gene Symbol: C5H14orf159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004941865.3 | XP_004941922.1 | 9 - 10 | 1724 | 88 | |
| XM_004941866.3 | XP_004941923.1 | 9 - 10 | 1489 | 88 | ||
| XM_015287745.2 | XP_015143231.1 | 9 - 10 | 1662 | 88 | ||
| XM_421319.6 | XP_421319.2 | 9 - 10 | 1409 | 88 | ||
| GenBank mRNA | BX933946.1 | - | 9 - 10 | 1323 | 88 | |
More Information
Set Membership: |
|

C5H14orf159
Genomic Map