Assay Details
| Gene Symbol: PBK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277999.1 | NP_001264928.1 | 5 - 6 | 609 | 58 | |
| XM_015285101.2 | XP_015140587.1 | 6 - 7 | 639 | 58 | ||
| XM_015285102.2 | XP_015140588.1 | 6 - 7 | 617 | 58 | ||
| GenBank mRNA | BU406153.1 | - | 2 - 3 | 184 | 58 | |
| BX930018.1 | - | 5 - 6 | 621 | 58 | ||
| BX934024.1 | - | 5 - 6 | 609 | 58 | ||
| CV860416.1 | - | 244 | 58 | |||
More Information
Set Membership: |
|

PBK
Genomic Map