Assay Details
| Gene Symbol: LIPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277754.1 | NP_001264683.1 | 2 | 1481 | 139 | |
| GenBank mRNA | BX929974.2 | - | 2 | 1481 | 139 | |
| BX934099.1 | - | 1 | 1153 | 139 | ||
More Information
Set Membership: |
|

LIPT1
Genomic Map