Assay Details
| Gene Symbol: STK32B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015285817.2 | XP_015141303.1 | 1 - 2 | 890 | 74 | |
| XM_420796.5 | XP_420796.2 | 1 - 2 | 885 | 74 | ||
| GenBank mRNA | BX934697.1 | - | 1 - 2 | 272 | 74 | |
More Information
Set Membership: |
|

STK32B
Genomic Map