Assay Details
| Gene Symbol: DRAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298810.2 | XP_015154296.1 | 481 | 66 | ||
| XM_015298811.2 | XP_015154297.1 | 636 | 66 | |||
| XM_025143844.1 | XP_024999612.1 | 447 | 66 | |||
| GenBank mRNA | BX932389.2 | - | 629 | 66 | ||
| BX935081.1 | - | 464 | 66 | |||
More Information
Set Membership: |
|

DRAM2
Genomic Map