Assay Details
| Gene Symbol: MRPS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277576.1 | NP_001264505.1 | 4 - 5 | 342 | 76 | |
| XM_015283680.2 | XP_015139166.1 | 5 - 6 | 401 | 76 | ||
| GenBank mRNA | BU262047.1 | - | 4 - 5 | 326 | 76 | |
| BX930192.1 | - | 4 - 5 | 329 | 76 | ||
| BX930686.1 | - | 4 - 5 | 315 | 76 | ||
| BX930708.2 | - | 4 - 5 | 319 | 76 | ||
| BX935171.1 | - | 5 - 6 | 518 | 76 | ||
| CD763841.1 | - | 4 - 5 | 331 | 76 | ||
| CF255365.1 | - | 4 - 5 | 344 | 76 | ||
| CR407465.1 | - | 4 - 5 | 310 | 76 | ||
More Information
Set Membership: |
|

MRPS10
Genomic Map