Assay Details
| Gene Symbol: SLC31A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277685.1 | NP_001264614.1 | 1 - 2 | 52 | 74 | |
| GenBank mRNA | BX935452.1 | - | 1 - 2 | 65 | 74 | |
| BX936102.1 | - | 1 - 2 | 52 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277685.1 transcripts.
Set Membership: |
|

SLC31A2
Genomic Map