Assay Details
| Gene Symbol: VPS29L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277505.1 | NP_001264434.1 | 3 - 4 | 466 | 101 | |
| XM_025155499.1 | XP_025011267.1 | 2 - 3 | 842 | 101 | ||
| GenBank mRNA | BX935605.1 | - | 3 - 4 | 467 | 101 | |
More Information
Set Membership: |
|

VPS29L
Genomic Map