Assay Details
| Gene Symbol: EMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414188.6 | XP_414188.1 | 1 - 2 | 315 | 66 | |
| GenBank mRNA | BX935865.1 | - | 1 - 2 | 270 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMC8| Gene Symbol: EMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414188.6 | XP_414188.1 | 1 - 2 | 315 | 66 | |
| GenBank mRNA | BX935865.1 | - | 1 - 2 | 270 | 66 | |
Set Membership: |
|