Assay Details
| Gene Symbol: MRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277545.1 | NP_001264474.1 | 2 - 3 | 318 | 81 | |
| XM_015294160.2 | XP_015149646.1 | 2 - 3 | 878 | 81 | ||
| XM_015294161.1 | XP_015149647.1 | 2 - 3 | 495 | 81 | ||
| XM_025155077.1 | XP_025010845.1 | 2 - 3 | 367 | 81 | ||
| GenBank mRNA | BX935892.1 | - | 2 - 3 | 319 | 81 | |
More Information
Set Membership: |
|

MRM2
Genomic Map