Assay Details
| Gene Symbol: MRAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320907.1 | NP_001307836.1 | 2 - 3 | 227 | 85 | |
| XM_015284715.2 | XP_015140201.1 | 6 - 7 | 765 | 85 | ||
| XM_025148762.1 | XP_025004530.1 | 5 - 6 | 1331 | 85 | ||
| XM_025148763.1 | XP_025004531.1 | 6 - 7 | 734 | 85 | ||
| XM_025148764.1 | XP_025004532.1 | 6 - 7 | 695 | 85 | ||
| XM_025148765.1 | XP_025004533.1 | 6 - 7 | 962 | 85 | ||
| XM_025148766.1 | XP_025004534.1 | 3 - 4 | 1121 | 85 | ||
| GenBank mRNA | BX929558.1 | - | 3 - 4 | 281 | 85 | |
| BX936128.1 | - | 6 - 7 | 757 | 85 | ||
| CR405797.1 | - | 4 - 5 | 447 | 85 | ||
| KT183012.1 | - | 2 - 3 | 227 | 85 | ||
More Information
Set Membership: |
|

MRAP2
Genomic Map