Assay Details
| Gene Symbol: EEF1E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418953.6 | XP_418953.5 | 3 - 4 | 472 | 140 | |
| GenBank mRNA | BX934973.2 | - | 3 - 4 | 425 | 140 | |
| BX950271.1 | - | 3 - 4 | 403 | 140 | ||
More Information
Set Membership: |
|

EEF1E1
Genomic Map