Assay Details
| Gene Symbol: THNSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936267.2 | XP_004936324.2 | 6 - 7 | 1072 | 84 | |
| XM_004936268.2 | XP_004936325.1 | 5 - 6 | 915 | 84 | ||
| XM_015285934.2 | XP_015141420.1 | 6 - 7 | 1329 | 84 | ||
| XM_015285935.2 | XP_015141421.1 | 6 - 7 | 1257 | 84 | ||
| XM_025150171.1 | XP_025005939.1 | 6 - 7 | 1426 | 84 | ||
| XM_420866.6 | XP_420866.4 | 6 - 7 | 1033 | 84 | ||
| GenBank mRNA | BX931386.1 | - | 2 - 3 | 285 | 84 | |
| BX931830.1 | - | 2 - 3 | 223 | 84 | ||
| BX950326.1 | - | 4 - 5 | 719 | 84 | ||
More Information
Set Membership: |
|

THNSL2
Genomic Map