Assay Details
| Gene Symbol: CTNS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015295844.2 | XP_015151330.1 | 8 - 9 | 751 | 86 | |
| XM_415851.6 | XP_415851.1 | 8 - 9 | 880 | 86 | ||
| GenBank mRNA | BX950718.1 | - | 8 - 9 | 927 | 86 | |
More Information
Set Membership: |
|

CTNS
Genomic Map