Assay Details
| Gene Symbol: CZH5ORF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145052.1 | XP_025000820.1 | 9 - 10 | 4299 | 131 | |
| XM_025145053.1 | XP_025000821.1 | 9 - 10 | 4299 | 131 | ||
| XM_025145054.1 | XP_025000822.1 | 9 - 10 | 4299 | 131 | ||
| XM_025145055.1 | XP_025000823.1 | 9 - 10 | 4299 | 131 | ||
| XM_025145056.1 | XP_025000824.1 | 9 - 10 | 4299 | 131 | ||
| GenBank mRNA | CR352435.1 | - | 4 - 5 | 395 | 131 | |
| CR386217.1 | - | 5 - 6 | 525 | 131 | ||
More Information
Set Membership: |
|

CZH5ORF34
Genomic Map