Assay Details
| Gene Symbol: OMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291181.2 | XP_015146667.1 | 5 - 6 | 1046 | 72 | |
| XM_422503.6 | XP_422503.3 | 5 - 6 | 1180 | 72 | ||
| GenBank mRNA | CR352620.1 | - | 4 - 5 | 876 | 72 | |
More Information
Set Membership: |
|

OMA1
Genomic Map