Assay Details
| Gene Symbol: SHROOM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155024.1 | XP_025010792.1 | 2 - 3 | 745 | 61 | |
| GenBank mRNA | CR352916.1 | - | 1 - 2 | 29 | 61 | |
| CR386996.1 | - | 2 - 3 | 229 | 61 | ||
More Information
Set Membership: |
|

SHROOM1
Genomic Map