Assay Details
| Gene Symbol: PYROXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277276.1 | NP_001264205.1 | 5 - 6 | 547 | 83 | |
| XM_025148920.1 | XP_025004688.1 | 2 - 3 | 199 | 83 | ||
| GenBank mRNA | BX929345.2 | - | 5 - 6 | 548 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025148920.1 transcripts.
Set Membership: |
|

PYROXD1
Genomic Map