Assay Details
| Gene Symbol: EIF4E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641762.4 | XP_003641810.1 | 3 - 4 | 336 | 73 | |
| XM_004943383.3 | XP_004943440.1 | 3 - 4 | 431 | 73 | ||
| XM_004943384.3 | XP_004943441.1 | 3 - 4 | 363 | 73 | ||
| XM_422748.5 | XP_422748.2 | 3 - 4 | 329 | 73 | ||
| GenBank mRNA | BX929435.2 | - | 3 - 4 | 295 | 73 | |
| BX929577.1 | - | 1 - 2 | 155 | 73 | ||
| BX931838.2 | - | 3 - 4 | 307 | 73 | ||
| CR354365.1 | - | 3 - 4 | 559 | 73 | ||
| CR385832.1 | - | 3 - 4 | 315 | 73 | ||
More Information
Set Membership: |
|

EIF4E2
Genomic Map