Assay Details
| Gene Symbol: DBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276354.1 | NP_001263283.1 | 2 - 3 | 483 | 64 | |
| XM_025147738.1 | XP_025003506.1 | 2 - 3 | 890 | 64 | ||
| GenBank mRNA | BX929917.2 | - | 2 - 3 | 242 | 64 | |
More Information
Set Membership: |
|

DBX2
Genomic Map