Assay Details
| Gene Symbol: DBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276354.1 | NP_001263283.1 | 3 - 4 | 682 | 78 | |
| GenBank mRNA | BX929917.2 | - | 3 - 4 | 441 | 78 | |
More Information
Set Membership: |
|

DBX2
Genomic Map