Assay Details
| Gene Symbol: NQO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418973.6 | XP_418973.1 | 3 - 4 | 816 | 61 | |
| GenBank mRNA | BX930070.2 | - | 3 - 4 | 574 | 61 | |
| BX932925.1 | - | 2 - 3 | 263 | 61 | ||
| BX934365.2 | - | 2 - 3 | 280 | 61 | ||
More Information
Set Membership: |
|

NQO2
Genomic Map