Assay Details
| Gene Symbol: C1H22orf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416268.6 | XP_416268.3 | 5 - 6 | 657 | 144 | |
| GenBank mRNA | BX930193.2 | - | 5 - 6 | 628 | 144 | |
| BX950503.2 | - | 5 - 6 | 642 | 144 | ||
More Information
Set Membership: |
|

C1H22orf23
Genomic Map