Assay Details
| Gene Symbol: STX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252158.1 | NP_001239087.1 | 5 - 6 | 515 | 64 | |
| XM_015285761.2 | XP_015141247.1 | 5 - 6 | 728 | 64 | ||
| GenBank mRNA | BX929347.1 | - | 5 - 6 | 504 | 64 | |
| BX930630.2 | - | 5 - 6 | 521 | 64 | ||
More Information
Set Membership: |
|

STX18
Genomic Map