Assay Details
| Gene Symbol: C1GALT1C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277789.1 | NP_001264718.1 | 2 | 202 | 72 | |
| XM_015278296.2 | XP_015133782.1 | 2 | 281 | 72 | ||
| GenBank mRNA | BX930690.2 | - | 1 | 144 | 72 | |
More Information
Set Membership: |
|

C1GALT1C1
Genomic Map