Assay Details
| Gene Symbol: CZH5ORF63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX930858.2 | - | 1 - 2 | 54 | 79 | |
| CR353733.1 | - | 1 - 2 | 57 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CZH5ORF63| Gene Symbol: CZH5ORF63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX930858.2 | - | 1 - 2 | 54 | 79 | |
| CR353733.1 | - | 1 - 2 | 57 | 79 | ||
Set Membership: |
|