Assay Details
| Gene Symbol: CZH5ORF63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015280530.1 | XP_015136016.1 | 4 - 5 | 78 | ||
| GenBank mRNA | BX930858.2 | - | 2 - 3 | 171 | 78 | |
| BX933223.2 | - | 3 - 4 | 275 | 78 | ||
| CR353733.1 | - | 2 - 3 | 78 | |||
More Information
Set Membership: |
|

CZH5ORF63
Genomic Map