Assay Details
| Gene Symbol: LRRC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277456.1 | NP_001264385.1 | 3 - 4 | 288 | 102 | |
| GenBank mRNA | BX932426.2 | - | 3 - 4 | 288 | 102 | |
More Information
Set Membership: |
|

LRRC6
Genomic Map