Assay Details
Gene Symbol: NSMCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277463.1 | NP_001264392.1 | 4 - 5 | 466 | 129 | |
NR_102407.1 | 3 - 4 | 359 | 129 | |||
GenBank mRNA | BX930919.2 | - | 4 - 5 | 469 | 129 | |
BX932130.1 | - | 3 - 4 | 359 | 129 | ||
BX932910.2 | - | 4 - 5 | 459 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() |
