Assay Details
| Gene Symbol: STAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282001.2 | XP_015137487.1 | 5 - 6 | 582 | 65 | |
| GenBank mRNA | BX933125.2 | - | 5 - 6 | 526 | 65 | |
More Information
Set Membership: |
|

STAM
Genomic Map