Assay Details
| Gene Symbol: GPR87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291847.2 | XP_015147333.1 | 1 - 2 | 455 | 86 | |
| GenBank mRNA | BX933590.2 | - | 2 - 3 | 301 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015291847.2 transcripts.
Set Membership: |
|

GPR87
Genomic Map