Assay Details
| Gene Symbol: GPR87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291847.2 | XP_015147333.1 | 2 - 3 | 585 | 149 | |
| GenBank mRNA | BX933590.2 | - | 3 - 4 | 431 | 149 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015291847.2 transcripts.
Set Membership: |
|

GPR87
Genomic Map