Assay Details
| Gene Symbol: PEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198642.1 | NP_001185571.1 | 3 - 4 | 404 | 78 | |
| GenBank mRNA | BX933434.2 | - | 3 - 4 | 365 | 78 | |
| BX933665.2 | - | 3 - 4 | 400 | 78 | ||
| BX934603.1 | - | 3 - 4 | 388 | 78 | ||
| CN222635.1 | - | 3 - 4 | 404 | 78 | ||
More Information
Set Membership: |
|

PEBP1
Genomic Map