Assay Details
| Gene Symbol: EGFL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305681.1 | NP_001292610.1 | 3 - 4 | 407 | 114 | |
| XM_025156056.1 | XP_025011824.1 | 3 - 4 | 274 | 114 | ||
| GenBank mRNA | BX932113.1 | - | 1 - 2 | 191 | 114 | |
| BX935022.2 | - | 1 - 2 | 186 | 114 | ||
More Information
Set Membership: |
|

EGFL7
Genomic Map