Assay Details
| Gene Symbol: TBL1X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159754.3 | NP_001153226.1 | 6 - 7 | 858 | 73 | |
| XM_015273683.2 | XP_015129169.1 | 8 - 9 | 1096 | 73 | ||
| XM_015273688.2 | XP_015129174.1 | 8 - 9 | 1062 | 73 | ||
| XM_015273692.2 | XP_015129178.1 | 7 - 8 | 1010 | 73 | ||
| XM_015273697.2 | XP_015129183.1 | 7 - 8 | 988 | 73 | ||
| XM_025150901.1 | XP_025006669.1 | 9 - 10 | 1137 | 73 | ||
| XM_025150902.1 | XP_025006670.1 | 7 - 8 | 1072 | 73 | ||
| XM_025150910.1 | XP_025006678.1 | 7 - 8 | 976 | 73 | ||
| GenBank mRNA | BX935206.2 | - | 7 - 8 | 922 | 73 | |
More Information
Set Membership: |
|

TBL1X
Genomic Map