Assay Details
| Gene Symbol: RPL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277728.1 | NP_001264657.1 | 2 - 3 | 206 | 60 | |
| GenBank mRNA | BU105412.1 | - | 1 - 2 | 155 | 60 | |
| BU222325.1 | - | 2 - 3 | 208 | 60 | ||
| BX935079.1 | - | 1 - 2 | 328 | 60 | ||
| BX935284.2 | - | 1 - 2 | 160 | 60 | ||
| CR386367.1 | - | 1 - 2 | 251 | 60 | ||
More Information
Set Membership: |
|

RPL8
Genomic Map