Assay Details
| Gene Symbol: HSPB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277613.1 | NP_001264542.1 | 2 - 3 | 274 | 86 | |
| GenBank mRNA | BU330715.1 | - | 2 - 3 | 274 | 86 | |
| BX933248.1 | - | 2 - 3 | 258 | 86 | ||
| BX935707.2 | - | 2 - 3 | 217 | 86 | ||
More Information
Set Membership: |
|

HSPB11
Genomic Map