Assay Details
| Gene Symbol: RAMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144973.1 | NP_001138445.1 | 3 - 4 | 214 | 111 | |
| XM_025147235.1 | XP_025003003.1 | 3 - 4 | 175 | 111 | ||
| GenBank mRNA | BX932709.1 | - | 3 - 4 | 265 | 111 | |
| BX935935.2 | - | 3 - 4 | 414 | 111 | ||
| FJ545727.1 | - | 3 - 4 | 214 | 111 | ||
More Information
Set Membership: |
|

RAMP3
Genomic Map