Assay Details
| Gene Symbol: CCNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414493.6 | XP_414493.1 | 2 - 3 | 444 | 83 | |
| GenBank mRNA | BX935990.2 | - | 2 - 3 | 360 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCNG1| Gene Symbol: CCNG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414493.6 | XP_414493.1 | 2 - 3 | 444 | 83 | |
| GenBank mRNA | BX935990.2 | - | 2 - 3 | 360 | 83 | |
Set Membership: |
|