Assay Details
| Gene Symbol: RAB11FIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421784.6 | XP_421784.1 | 1 - 2 | 535 | 90 | |
| GenBank mRNA | CR353328.1 | - | 1 - 2 | 527 | 90 | |
More Information
Set Membership: |
|

RAB11FIP2
Genomic Map