Assay Details
| Gene Symbol: RPS27L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277460.1 | NP_001264389.1 | 3 - 4 | 344 | 93 | |
| GenBank mRNA | BX260540.3 | - | 3 - 4 | 344 | 93 | |
| BX276660.2 | - | 2 - 3 | 300 | 93 | ||
| BX934988.1 | - | 3 - 4 | 244 | 93 | ||
| CR354006.1 | - | 3 - 4 | 306 | 93 | ||
More Information
Set Membership: |
|

RPS27L
Genomic Map