Assay Details
| Gene Symbol: PRPF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937491.3 | XP_004937548.1 | 10 - 11 | 74 | ||
| XM_417229.6 | XP_417229.1 | 9 - 10 | 74 | |||
| GenBank mRNA | CR354060.1 | - | 3 - 4 | 331 | 74 | |
More Information
Set Membership: |
|

PRPF18
Genomic Map