Assay Details
| Gene Symbol: KCNMB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199630.1 | NP_001186559.1 | 1 - 2 | 353 | 123 | |
| GenBank mRNA | BX931406.2 | - | 1 - 2 | 353 | 123 | |
| CR354309.1 | - | 1 - 2 | 297 | 123 | ||
More Information
Set Membership: |
|

KCNMB4
Genomic Map