Assay Details
| Gene Symbol: CENPW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277733.1 | NP_001264662.1 | 1 - 2 | 176 | 64 | |
| XM_015284433.2 | XP_015139919.1 | 1 - 2 | 196 | 64 | ||
| GenBank mRNA | CR385186.1 | - | 1 - 2 | 176 | 64 | |
More Information
Set Membership: |
|

CENPW
Genomic Map