Assay Details
| Gene Symbol: PRDM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004941559.3 | XP_004941616.1 | 4 - 5 | 392 | 100 | |
| XM_015287188.2 | XP_015142674.1 | 6 - 7 | 758 | 100 | ||
| XM_015287189.2 | XP_015142675.1 | 6 - 7 | 1231 | 100 | ||
| XM_015287191.2 | XP_015142677.1 | 3 - 4 | 310 | 100 | ||
| XM_015287192.2 | XP_015142678.1 | 3 - 4 | 288 | 100 | ||
| XM_421099.6 | XP_421099.3 | 5 - 6 | 685 | 100 | ||
| GenBank mRNA | CR385630.1 | - | 5 - 6 | 685 | 100 | |
More Information
Set Membership: |
|

PRDM11
Genomic Map