Assay Details
| Gene Symbol: LRRC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418756.5 | XP_418756.1 | 1 - 2 | 297 | 89 | |
| GenBank mRNA | CR386255.1 | - | 1 - 2 | 88 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_418756.5 transcripts.
Set Membership: |
|

LRRC3B
Genomic Map