Assay Details
| Gene Symbol: POLR2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293302.1 | NP_001280231.1 | 6 - 7 | 474 | 62 | |
| GenBank mRNA | BX929268.1 | - | 6 - 7 | 476 | 62 | |
| BX930348.2 | - | 6 - 7 | 438 | 62 | ||
| CR386275.1 | - | 6 - 7 | 441 | 62 | ||
More Information
Set Membership: |
|

POLR2C
Genomic Map